Learn More
Medchemexpress LLC 4-methylphenyl 1-thio-β-D-glucopyranoside | 1152-39-2 | MFCD11045365 | 99.0% | 286.34 g/mol | C13H18O5S | 5 G
SDP
Catalog No. 5000285158
Encompass_Preferred
4-Methylphenyl 1-thio-β-D-glucopyranoside is a thioglycoside reagent for glycobiology research, commonly used as a substrate in enzymatic assays and studies of glycan recognition. Supplied as a purified white crystalline solid for research and analytical applications (not for human use).
  • Used as a substrate for glycosidase and glycosyltransferase assays.
  • High purity suitable for analytical and biochemical research.
  • White crystalline solid with molecular formula C13H18O5S.
  • CAS number 1152-39-2 for unambiguous identification.
  • Available in multiple pack sizes including 5 G for lab-scale experiments.
Catalog No. 50-002-85158 Supplier Medchemexpress LLC Supplier No. HYW3571255G
4-Methylphenyl 1-thio-β-D-glucopyranoside is a thioglycoside reagent for glycobiology research, commonly used as a substrate in enzymatic assays and studies of glycan recognition. Supplied as a purified white crystalline solid for research and analytical applications (not for human use).
  • Used as a substrate for glycosidase and glycosyltransferase assays.
  • High purity suitable for analytical and biochemical research.
  • White crystalline solid with molecular formula C13H18O5S.
  • CAS number 1152-39-2 for unambiguous identification.
  • Available in multiple pack sizes including 5 G for lab-scale experiments.