Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylphthalic Acid, Spectrum™ Chemical
SDP

Catalog No. 18621023 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-023 25 g
18-621-024 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-621-023 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M144425GM
Add to Cart
Add to Cart

Synonym: 4-Methyl-Phthalic Acid, 4-Methyl-o-Phthalate, 4-Methyl-1,2-Benzenedicarboxylic Acid

Chemical Identifiers

CAS 4316-23-8
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.16
InChI Key CWJJAFQCTXFSTA-UHFFFAOYSA-N
IUPAC Name 4-methylbenzene-1,2-dicarboxylic acid
SMILES CC1=CC=C(C(O)=O)C(=C1)C(O)=O

Specifications

Packaging Amber Bottle
Quantity 25 g
Percent Purity 98%
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.