Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methylphthalimide | 40314-06-5 | MFCD05155224 | 100.0% | 161.16 g/mol | C9H7NO2 | 5 G
SDP

Catalog No. 5000210011
Encompass_Preferred

4-Methylphthalimide is a fluorescent stain used in biological research applications, supplied as a solid reagent for laboratory use.

  • Stain for biological research applications.
  • Molecular formula C9H7NO2; molecular weight 161.16 g/mol.
  • CAS number 40314-06-5 for unambiguous identification.
  • High purity (99.96% as reported by supplier).
  • Appearance: off-white to light brown solid.
  • Documentation available: data sheet, COA, and safety data sheet (SDS).

Catalog No. 50-002-10011 Supplier Medchemexpress LLC Supplier No. HYW0538715G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-Methylphthalimide is a fluorescent stain used in biological research applications, supplied as a solid reagent for laboratory use.

  • Stain for biological research applications.
  • Molecular formula C9H7NO2; molecular weight 161.16 g/mol.
  • CAS number 40314-06-5 for unambiguous identification.
  • High purity (99.96% as reported by supplier).
  • Appearance: off-white to light brown solid.
  • Documentation available: data sheet, COA, and safety data sheet (SDS).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.