Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-methylpiperazine-2,2,6,6-d4 | 343864-02-8 | 99.4% | 104.19 g/mol | C5H8D4N2 | 10 MG
SDP

Catalog No. 5000298773
Encompass_Preferred

N-Methylpiperazine-d4 is the deuterium-labeled analogue of N-methylpiperazine, provided as a research-grade internal standard and tracer for analytical and isotope-labeling studies. It is designed for quantitative analysis by NMR, GC-MS, and LC-MS.

  • Deuterium-labeled (d4) compound for tracer studies.
  • Molecular formula C5H8D4N2.
  • Molecular weight 104.19 g/mol.
  • High purity (99.4%) suitable for analytical use.
  • Appearance: liquid, colorless to light yellow.
  • Recommended storage: pure form at -20°C.

Catalog No. 50-002-98773 Supplier Medchemexpress LLC Supplier No. HY78871S210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Methylpiperazine-d4 is the deuterium-labeled analogue of N-methylpiperazine, provided as a research-grade internal standard and tracer for analytical and isotope-labeling studies. It is designed for quantitative analysis by NMR, GC-MS, and LC-MS.

  • Deuterium-labeled (d4) compound for tracer studies.
  • Molecular formula C5H8D4N2.
  • Molecular weight 104.19 g/mol.
  • High purity (99.4%) suitable for analytical use.
  • Appearance: liquid, colorless to light yellow.
  • Recommended storage: pure form at -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.