Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Methylpiperidine solution

Catalog No. 792152500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mL
2 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-215-2500ML 500 mL
79-215-24L 4 L
79-215-22L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 79-215-2500ML Supplier Sigma Aldrich Supplier No. 792152500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-methylpiperidine solution (20 % in dmf) can be used:
  • In the deprotection of f-moc protecting group which is attached to free amine
  • In the determination of resin-bound amino acid by UV-fmoc-quantitation

Specifications

CAS 626-58-4
Density 0.920 g/mL (at 25°C)
Molecular Formula C6H13N
Refractive Index n20/D 1.433
Linear Formula C6H13N
MDL Number MFCD00006005
Quantity 500 mL
Synonym 4-Methylpiperidine solution
Molecular Weight (g/mol) 99.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.