Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylpyridazine, 99%, Thermo Scientific™

Catalog No. AAL09749ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL09749ME 500 mg
AAL09749MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL09749ME Supplier Thermo Scientific Chemicals Supplier No. L09749ME

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-88-3
Molecular Formula C5H6N2
Molecular Weight (g/mol) 94.117
MDL Number MFCD00011594
InChI Key AIKUBOPKWKZULG-UHFFFAOYSA-N
Synonym pyridazine, 4-methyl, 4-methyl pyridazine, 4-methyl-1,2-diazine, 4-methylpyridazine, acmc-209z0j, ksc183s2h, 1,2-oxathiolane 2,2-dioxide; 3-hydroxy-1-propanesulfonic acid gamma-sultone; propane sultone
PubChem CID 136882
IUPAC Name 4-methylpyridazine
SMILES CC1=CN=NC=C1

Specifications

Density 1.06
Boiling Point 98°C to 100°C (11 mmHg)
Flash Point 105°C (221°F)
Refractive Index 1.521
Quantity 500 mg
Beilstein 105689
Formula Weight 94.12
Percent Purity 99%
Chemical Name or Material 4-Methylpyridazine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 620-972-5
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.