Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Methylstyrene

Catalog No. 503124394 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-4394 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-312-4394 Supplier Sigma Aldrich Supplier No. M80806100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Methylstyrene is a molecule with an extended +ß conjugation. The threefold symmetric torsional potential of 4-methylstyrene has been investigated. Polymerization of 4-methylstyrene by employing a half-metallocene type catalytic system composed of (trimethyl)pentamethylcyclopentadienyltitanium (CpTiMe3), trioctylaluminum (AlOct3), and tris(pentafluorophenyl)borane [B(C6F5)3] has been reported. Palladium-catalyzed Heck coupling of chlorobenzene with 4-methylstyrene has been investigated.4-Methylstyrene was employed as +ß ligand in the preparation of cationic, two-coordinate triphenylphosphine-gold(I)-+ß complexes.

Specifications

CAS 622-97-9
Density 0.897 g/mL (at 25°C)
Boiling Point 170°C to 175°C
Molecular Formula C9H10
Linear Formula CH3C6H4CH=CH2
Refractive Index n20/D 1.542 (literature)
Quantity 100 mL
MDL Number MFCD00008621
Synonym 4-Vinyltoluene
Molecular Weight (g/mol) 118.18
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.