Learn More
Medchemexpress LLC 4-(Methylsulfonyl)phenylboronic acid | 149104-88-1 | 99.8% | C7H9BO4S | 10 G
SDP
Catalog No. 5000433459
Encompass_Preferred
4-(Methylsulfonyl)phenylboronic acid is a chemical intermediate. It is utilized in the synthesis of GPR119 agonists through the Suzuki coupling reaction. This compound also finds application in the research of type 2 diabetes.
  • Used as a chemical intermediate
  • Synthesizes GPR119 agonists via Suzuki coupling reaction
  • Applicable in type 2 diabetes research
Catalog No. 50-004-33459 Supplier Medchemexpress LLC Supplier No. HY7764210G
Add to Cart
Add to Cart
4-(Methylsulfonyl)phenylboronic acid is a chemical intermediate. It is utilized in the synthesis of GPR119 agonists through the Suzuki coupling reaction. This compound also finds application in the research of type 2 diabetes.
  • Used as a chemical intermediate
  • Synthesizes GPR119 agonists via Suzuki coupling reaction
  • Applicable in type 2 diabetes research