Learn More
Medchemexpress LLC 4-(Methylsulfonyl)phenylboronic acid | 149104-88-1 | 99.84% | C7H9BO4S | 5 G
SDP
Catalog No. 5000434966
Encompass_Preferred
4-(Methylsulfonyl)phenylboronic acid is an intermediate compound primarily used in the synthesis of GPR119 agonists through Suzuki coupling reactions. This chemical is also valuable for research related to type 2 diabetes. It presents as a white to off-white solid and is exclusively for research purposes, not for medical applications or patient use.
  • Intermediate for chemical synthesis
  • Synthesizes GPR119 agonists via Suzuki coupling reaction
  • Used in type 2 diabetes research
  • Appears as a white to off-white solid
  • Intended for research use only
Catalog No. 50-004-34966 Supplier Medchemexpress LLC Supplier No. HY776425G
Add to Cart
Add to Cart
4-(Methylsulfonyl)phenylboronic acid is an intermediate compound primarily used in the synthesis of GPR119 agonists through Suzuki coupling reactions. This chemical is also valuable for research related to type 2 diabetes. It presents as a white to off-white solid and is exclusively for research purposes, not for medical applications or patient use.
  • Intermediate for chemical synthesis
  • Synthesizes GPR119 agonists via Suzuki coupling reaction
  • Used in type 2 diabetes research
  • Appears as a white to off-white solid
  • Intended for research use only