Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylthiazole-5-carboxylic acid, 97%, Thermo Scientific™

Catalog No. AC396580250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC396580250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC396580250 Supplier Thermo Scientific Chemicals Supplier No. 396580250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20485-41-0
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.17
InChI Key ZGWGSEUMABQEMD-UHFFFAOYSA-N
Synonym 4-methylthiazole-5-carboxylic acid, 4-methyl-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 4-methyl, 4-methyl-thiazole-5-carboxylic acid, 4-methyl-5-thiazole carboxylic acid, 4-methylthiazole-5-carboxylicacid, pubchem8416, acmc-209far, 4-27-00-04008 beilstein handbook reference, ksc201q4f
PubChem CID 209805
IUPAC Name 4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC=N1)C(=O)O

Specifications

Melting Point 287.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 143.17
Percent Purity 97%
Chemical Name or Material 4-Methylthiazole-5-carboxylic acid
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.