Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylthiazole-5-carboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. M22355G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22355G Supplier TCI America Supplier No. M22355G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 20485-41-0
Molecular Formula C5H5NO2S
Molecular Weight (g/mol) 143.16
MDL Number MFCD00626872
InChI Key ZGWGSEUMABQEMD-UHFFFAOYSA-N
Synonym 4-methylthiazole-5-carboxylic acid, 4-methyl-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 4-methyl, 4-methyl-thiazole-5-carboxylic acid, 4-methyl-5-thiazole carboxylic acid, 4-methylthiazole-5-carboxylicacid, pubchem8416, acmc-209far, 4-27-00-04008 beilstein handbook reference, ksc201q4f
PubChem CID 209805
IUPAC Name 4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC=N1)C(=O)O

Specifications

Melting Point 260°C
Quantity 5 g
Formula Weight 143.16
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylthiazole-5-carboxylic Acid
RTECSNumber XJ3680000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.