Learn More
Sigma Aldrich Fine Chemicals Biosciences 4 Methylthiazole 98 FG1KG
Catalog No. 502908328
Encompass
4-Methylthiazole is one of the key volatile flavor products formed during the thermal degradation of thiamin. It is reported to be found in roasted peanuts and roasted beef.
Catalog No. 50-290-8328 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W3716021KGK
4-Methylthiazole is one of the key volatile flavor products formed during the thermal degradation of thiamin. It is reported to be found in roasted peanuts and roasted beef.