Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(Methylthio)benzaldehyde, 97%

Click to view available options
Quantity:
25g
100g
3446-89-7
C8H8OS
152.211
MFCD00006948
QRVYABWJVXXOTN-UHFFFAOYSA-N
4-methylthio benzaldehyde,benzaldehyde, 4-methylthio,4-methylsulfanyl benzaldehyde,4-methylmercaptobenzaldehyde,p-methylthio benzaldehyde,p-methylthiobenzaldehyde,unii-1tvq11biy8,4-aldehyde thioanisole,p-methylmercaptobenzaldehyde
76985
4-methylsulfanylbenzaldehyde
CSC1=CC=C(C=C1)C=O
3446-89-7
C8H8OS
152.211
MFCD00006948
QRVYABWJVXXOTN-UHFFFAOYSA-N
4-methylthio benzaldehyde,benzaldehyde, 4-methylthio,4-methylsulfanyl benzaldehyde,4-methylmercaptobenzaldehyde,p-methylthio benzaldehyde,p-methylthiobenzaldehyde,unii-1tvq11biy8,4-aldehyde thioanisole,p-methylmercaptobenzaldehyde
76985
4-methylsulfanylbenzaldehyde
CSC1=CC=C(C=C1)C=O
Applications
4-(Methylthio)benzaldehyde is a building block for the synthesis of various pharmaceutical and biologically active compounds. It can be used for the synthesis of sulfur-containing terpyridine ligands. It is the intermediate for the synthesis of a class of pyrrole derivatives showing analgesic/anti-inflammatory activity.

Solubility
Not miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
Flash Point >110°C (230°F)
Boiling Point 118°C to 121°C (5mmHg)
Melting Point 6°C
Quantity 25g
Beilstein 636158
Formula Weight 152.22
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)benzaldehyde