Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylthio)benzaldehyde, 97%

Catalog No. p-168382
Change view
Click to view available options
Quantity:
5 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207150050 5 mL Glass bottle
AC207150500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC207150050 Supplier Thermo Scientific Chemicals Supplier No. 207150050
Only null left
Add to Cart

CAS: 3446-89-7 | C8H8OS | 152.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3446-89-7
Molecular Formula C8H8OS
Molecular Weight (g/mol) 152.21
MDL Number MFCD00006948
InChI Key QRVYABWJVXXOTN-UHFFFAOYSA-N
Synonym 4-methylthio benzaldehyde, benzaldehyde, 4-methylthio, 4-methylsulfanyl benzaldehyde, 4-methylmercaptobenzaldehyde, p-methylthio benzaldehyde, p-methylthiobenzaldehyde, unii-1tvq11biy8, 4-aldehyde thioanisole, p-methylmercaptobenzaldehyde
PubChem CID 76985
IUPAC Name 4-methylsulfanylbenzaldehyde
SMILES CSC1=CC=C(C=C1)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 6.0°C
Color Yellow
Density 1.1400g/mL
Boiling Point 86.0°C to 90.0°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.6452
Linear Formula CH3SC6H4CHO
Quantity 5 mL
Beilstein 08, I, 533
Specific Gravity 1.14
Formula Weight 152.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(Methylthio)benzaldehyde, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
EINECSNumber 222-365-7
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.