Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylthio)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. M14581G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14581G 1 g
M1458-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M14581G Supplier TCI America Supplier No. M14581G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material 4-(Methylthio)phenylboronic Acid (contains varying amounts of Anhydride)
CAS 98546-51-1
Melting Point 213°C
Color White-Yellow
Molecular Formula C7H9BO2S
Quantity 1 g
MDL Number MFCD00093410
Synonym 4-methylthio phenylboronic acid, 4-thioanisoleboronic acid, 4-methylsulfanyl phenylboronic acid, 4-methylthiophenylboronic acid, 4-methylthio phenyl boronic acid, 4-methylsulfanylphenyl boronic acid, 4-methylthio phenylboronicacid, 4-methylthio benzeneboronic acid, thioanisole-4-boronic acid, 4-methylmercapto benzeneboronic acid
InChI Key IVUHTLFKBDDICS-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)SC)(O)O
IUPAC Name (4-methylsulfanylphenyl)boronic acid
Molecular Weight (g/mol) 168.017
PubChem CID 2733959
Formula Weight 168.02
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.