Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylthiomorpholine 1,1-dioxide, 97%, Thermo Scientific™

Catalog No. AAL0223503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0223503 1 g
AAL0223506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0223503 Supplier Thermo Scientific Chemicals Supplier No. L0223503

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25343-91-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.208
MDL Number MFCD00014623
InChI Key ULZCOWMSBOJCLT-UHFFFAOYSA-N
PubChem CID 91413
IUPAC Name 4-methyl-1,4-thiazinane 1,1-dioxide
SMILES CN1CCS(=O)(=O)CC1

Specifications

Melting Point 84°C to 88°C
Quantity 1 g
Beilstein 114527
Synonym 4-methylthiomorpholine 1,1-dioxide, 4-methylthiomorpholine-1,1-dioxide, thiomorpholine, 4-methyl-, 1,1-dioxide, n-methyltetrahydro-1,4-thiazine-s-oxide, 4-methyl-1??-thiomorpholine-1,1-dione, 4-methylthiomorpholine1,1-dioxide, acmc-1cc0z, wln: t6n dswtj a1, 4-27-00-00636 beilstein handbook reference, thiomorpholine,4-methyl-, 1,1-dioxide
Formula Weight 149.21
Percent Purity 97%
Chemical Name or Material 4-Methylthiomorpholine 1,1-dioxide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-877-5
RTECSNumber XM3015000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.