Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylthiosemicarbazide 95.0+%, TCI America™
SDP

Catalog No. M028925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M028925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M028925G Supplier TCI America Supplier No. M028925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6610-29-3
Molecular Formula C2H7N3S
Molecular Weight (g/mol) 105.16
MDL Number MFCD00007617
InChI Key PTVZQOAHCSKAAS-UHFFFAOYSA-N
Synonym 4-methylthiosemicarbazide, 4-methyl-3-thiosemicarbazide, hydrazinecarbothioamide, n-methyl, n-methylhydrazinecarbothioamide, methylthiosemicarbazide, 1-methyl-2-thiosemicarbazide, semicarbazide, 4-methyl-3-thio, n-methylthiosemicarbazide, 4-methylhydrazinecarbothioamide, unii-pk83e1t776
PubChem CID 2723853
IUPAC Name 3-amino-1-methylthiourea
SMILES CNC(=S)NN

Specifications

Melting Point 136°C
Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 105.16
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylthiosemicarbazide
EINECSNumber (9)-1443
RTECSNumber MV1407370
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.