Learn More
Cayman Chemical 4-Methylumbelliferone, Purity: ≥98%, CAS: 90-33-5, 500 g
SDP
Catalog No. 502990406
Encompass_Preferred
A synthetic coumarin with diverse biological activities; inhibits hyaluronic acid synthesis in PC3-ML and DU145 cells (IC50s = ~0.4 mM for both); inhibits the growth of PC3-ML, DU145, C4-2B, LNCaP, and LAPC-4 prostate cancer cells (IC50s = 0.2-0.4 mM) and invasion and chemotactic motility of PC3-ML and DU145 cells in vitro; reduces tumor growth and microvessel density in a PC3-ML mouse xenograft model at 225 and 450 mg/kg twice per day; active against H. pylori and V. cholerae in vitro (MICs = 59 and 100-200 μg/ml, respectively); ex/em = 320 and 360 nm at low and high pH, respectively/455 and 445 nm at low and high pH, respectively.
Catalog No. 50-299-0406 Supplier Cayman Chemical Supplier No. 29603500 G
A synthetic coumarin with diverse biological activities; inhibits hyaluronic acid synthesis in PC3-ML and DU145 cells (IC50s = ~0.4 mM for both); inhibits the growth of PC3-ML, DU145, C4-2B, LNCaP, and LAPC-4 prostate cancer cells (IC50s = 0.2-0.4 mM) and invasion and chemotactic motility of PC3-ML and DU145 cells in vitro; reduces tumor growth and microvessel density in a PC3-ML mouse xenograft model at 225 and 450 mg/kg twice per day; active against H. pylori and V. cholerae in vitro (MICs = 59 and 100-200 μg/ml, respectively); ex/em = 320 and 360 nm at low and high pH, respectively/455 and 445 nm at low and high pH, respectively.