Learn More
Cayman Chemical 4-Methylumbelliferyl-β-D-Galactoside, Purity: ≥98%, CAS: 6160-78-7, 1 g
SDP
Catalog No. 502946567
Encompass_Preferred
A fluorogenic substrate for β-galactosidase; hydrolysis of MUG by β-gal releases the fluorescent moiety 4-MU, which displays a pH-dependent excitation maximum of 330, 370, and 385 nm at pH 4.6, 7.4, and 10.4, respectively, and an emission maximum between 445-454 nm; has been used to detect β-gal activity in intact bacteria, yeast, and mammalian cells
Catalog No. 50-294-6567 Supplier Cayman Chemical Supplier No. 339201 G
A fluorogenic substrate for β-galactosidase; hydrolysis of MUG by β-gal releases the fluorescent moiety 4-MU, which displays a pH-dependent excitation maximum of 330, 370, and 385 nm at pH 4.6, 7.4, and 10.4, respectively, and an emission maximum between 445-454 nm; has been used to detect β-gal activity in intact bacteria, yeast, and mammalian cells