Learn More
Cayman Chemical 4-MethylumbelIferylD-Gluc 1g
SDP
Supplier:  Cayman Chemical 209481 G
Encompass_Preferred
A fluorogenic substrate of β-glucosidase and β-glucocerebrosidase
Catalog No. 50-298-3860