Learn More
Cayman Chemical 4-MethylumbelIferylD-Gluc 1g
SDP
Catalog No. 502966452
Encompass_Preferred
A fluorogenic substrate of β-glucuronidase; cleaved by β-glucuronide to release the fluorescent moiety 4-MU
Catalog No. 50-296-6452 Supplier Cayman Chemical Supplier No. 172031 G
A fluorogenic substrate of β-glucuronidase; cleaved by β-glucuronide to release the fluorescent moiety 4-MU