Learn More
Cayman Chemical 4-Methylumbelliferyl-β-D-Glucopyranoside, Purity: ≥98%, CAS: 18997-57-4, 5 g
SDP
Catalog No. 502983298
Encompass_Preferred
A fluorogenic substrate of β-glucosidase and β-glucocerebrosidase
Catalog No. 50-298-3298 Supplier Cayman Chemical Supplier No. 209485 G
A fluorogenic substrate of β-glucosidase and β-glucocerebrosidase