Learn More
Cayman Chemical 4-MethylumbelIferylD-Gluc 5g
SDP
Catalog No. 502965195
Encompass_Preferred
A fluorogenic substrate of β-glucuronidase; cleaved by β-glucuronide to release the fluorescent moiety 4-MU
Catalog No. 50-296-5195 Supplier Cayman Chemical Supplier No. 172035 G
A fluorogenic substrate of β-glucuronidase; cleaved by β-glucuronide to release the fluorescent moiety 4-MU