Learn More
Selleck Chemical LLC 4-Methylumbelliferone (4-MU) 10mg 90-33-5
SDP
Supplier:  Selleck Chemical LLC S225610MG
Encompass_Preferred
4-Methylumbelliferone is a hyaluronic acid (HA) synthesis inhibitor with an IC50 of 0.4 mM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-549-6