Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Methylumbelliferone

Catalog No. M138125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M138125G 25 g
M1381100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M138125G Supplier Sigma Aldrich Supplier No. M138125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 4-methylumbelliferone is primarily used in synthesizing medicinal compounds and as a building block for fluorescent probes. applications include synthesis of:
  • Coumarin triazole derivatives as potential antimicrobial agents
  • Coumarin salen-based fluorescence sensors for mg2+ detection
  • Pyranocoumarin derivatives as anti-hyperglycemic and anti-dyslipidemic agents
  • Coumarin piperazine derivatives as potential multireceptor atypical antipsychotics
  • 4-methylumbelliferyl t-antigen as a substrate for endo-α-n-acetylgalactosaminidase

Specifications

CAS 90-33-5
Molecular Formula C10H8O3
Linear Formula C10H8O3
MDL Number MFCD00006866
Quantity 25 g
Synonym beta-Methylumbelliferone; 4-MU; 7-Hydroxy-4-methylcoumarin
Molecular Weight (g/mol) 176.17
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.