Learn More
Research Products International Corp 4-Methylumbelliferyl-a-D-Galactopyranoside Monohydrate [4-Methylumbelliferyl-a-D-Galactoside], 1 Gram
SDP
Supplier:  Research Products International Corp M654001.0
Encompass_Preferred
Fluorescent substrate for α-D-Galactosidase.
Catalog No. 50-488-819