Learn More
Research Products International Corp 4-Methylumbelliferyl Elaidate, 100 Milligrams
SDP
Catalog No. 50213466
Encompass_Preferred
Fluorescent substrate for Esterase. Purity: > 98%. 4-Methylumbelliferone < 50ppm. Soluble in 5% toluene.
Catalog No. 50-213-466 Supplier Research Products International Corp Supplier No. M641650.1
Fluorescent substrate for Esterase. Purity: > 98%. 4-Methylumbelliferone < 50ppm. Soluble in 5% toluene.