Learn More
Research Products International Corp 4-Methylumbelliferyl-B-L-Fucopyranoside, 100 Milligrams
SDP
Catalog No. 50213469
Encompass_Preferred
Fluorescent substrate for β-L-Fucosidase.
  • 4-Methylumbelliferone <50ppm
Catalog No. 50-213-469 Supplier Research Products International Corp Supplier No. M651000.1
Fluorescent substrate for β-L-Fucosidase.
  • 4-Methylumbelliferone <50ppm