Learn More
Medchemexpress LLC 4-Methylumbelliferyl 10mM | 440.61 g/mol | C28H40O4 | 10MM 1ML
SDP
Catalog No. 5000229242
Encompass_Preferred
4-Methylumbelliferyl oleate is a fluorogenic substrate for acid and alkaline lipases 4-Methylumbelliferyl oleate is cleaved by lipases liberating 4-Methylumbelliferyl (Ex/Em 320/450 nm)[1][2]
Catalog No. 50-002-29242 Supplier Medchemexpress LLC Supplier No. HY11709510MM 1ML
4-Methylumbelliferyl oleate is a fluorogenic substrate for acid and alkaline lipases 4-Methylumbelliferyl oleate is cleaved by lipases liberating 4-Methylumbelliferyl (Ex/Em 320/450 nm)[1][2]