Learn More
Medchemexpress LLC 4-Methylumbelliferyl | 124223-99-0 | 10MG
SDP
Catalog No. 5000235522
Encompass_Preferred
4-Methylumbelliferyl 2-acetamido-2-deoxy- -D-galactopyranoside is a fluorogenic substrate for studying the enzymatic activity of -galactosidase 4-Methylumbelliferyl 2-acetamido-2-deoxy- -D-galactopyranoside is used to analyze enzyme activity in various biological samples helping scientists gain insight into the function of the enzyme in vivo 4-Methylumbelliferyl 2-acetamido-2-deoxy- -D-galactopyranoside can also be used to screen potential compounds to inhibit or activate this enzyme
Catalog No. 50-002-35522 Supplier Medchemexpress LLC Supplier No. HY12300010MG
4-Methylumbelliferyl 2-acetamido-2-deoxy- -D-galactopyranoside is a fluorogenic substrate for studying the enzymatic activity of -galactosidase 4-Methylumbelliferyl 2-acetamido-2-deoxy- -D-galactopyranoside is used to analyze enzyme activity in various biological samples helping scientists gain insight into the function of the enzyme in vivo 4-Methylumbelliferyl 2-acetamido-2-deoxy- -D-galactopyranoside can also be used to screen potential compounds to inhibit or activate this enzyme