Learn More
Medchemexpress LLC 4-Methylumbelliferyl | 124223-99-0 | MFCD00270047 | 99.0% | 379.36 g/mol | C18H21NO8 | 1MG
SDP
Catalog No. 5000238784
Encompass_Preferred
4-Methylumbelliferyl 2-acetamido-2-deoxy-α-D-galactopyranoside is a fluorogenic glycoside substrate that liberates 4-methylumbelliferone upon enzymatic hydrolysis, enabling sensitive fluorescence-based detection of glycosidase activity in biochemical assays.
  • Releases a fluorescent reporter for sensitive enzyme assays
  • High purity (99.0%) for consistent assay performance
  • White to off-white solid, easy to handle and dissolve
  • Suitable for measuring glycosidase activity in biochemical assays
  • Stable under recommended storage for long-term use
Catalog No. 50-002-38784 Supplier Medchemexpress LLC Supplier No. HY1230001MG
4-Methylumbelliferyl 2-acetamido-2-deoxy-α-D-galactopyranoside is a fluorogenic glycoside substrate that liberates 4-methylumbelliferone upon enzymatic hydrolysis, enabling sensitive fluorescence-based detection of glycosidase activity in biochemical assays.
  • Releases a fluorescent reporter for sensitive enzyme assays
  • High purity (99.0%) for consistent assay performance
  • White to off-white solid, easy to handle and dissolve
  • Suitable for measuring glycosidase activity in biochemical assays
  • Stable under recommended storage for long-term use