Learn More
Medchemexpress LLC 4-Methylumbelliferyl 1g
SDP
Catalog No. 5000205587
Encompass_Preferred
4-Methylumbelliferyl octanoate is a octoate 4-Methylumbelliferyl octanoate can be used as a s ubstrate of hog pancreatic lipase[1]
Catalog No. 50-002-05587 Supplier Medchemexpress LLC Supplier No. HYW0996481G
4-Methylumbelliferyl octanoate is a octoate 4-Methylumbelliferyl octanoate can be used as a s ubstrate of hog pancreatic lipase[1]