Learn More
Alkali Scientific 4 Methylumbelliferyl 2 acetamido 2 deoxy β D galactopyranoside [4 Methylumbelliferyl N acetyl β D galactosaminide], 25 Mg
SDP
Catalog No. 5000660548
Encompass_Preferred
Methylumbelliferyl β-D-Galactosaminide (25mg) is a high-purity biochemical reagent used in assays to analyze enzymatic activity. This compound is commonly employed in research to study enzymes that hydrolyze galactosamine-containing substrates, such as galactosaminidases. Upon enzymatic hydrolysis, the substrate produces a fluorescent product, making it ideal for high-sensitivity detection of enzyme activity. The 25mg product is suitable for laboratory-scale experiments and provides a reliable tool for studying carbohydrate metabolism, enzyme kinetics, and substrate specificity in biochemical and pharmaceutical research.
Catalog No. 50-006-60548 Supplier Alkali Scientific Supplier No. LLI001CP
Methylumbelliferyl β-D-Galactosaminide (25mg) is a high-purity biochemical reagent used in assays to analyze enzymatic activity. This compound is commonly employed in research to study enzymes that hydrolyze galactosamine-containing substrates, such as galactosaminidases. Upon enzymatic hydrolysis, the substrate produces a fluorescent product, making it ideal for high-sensitivity detection of enzyme activity. The 25mg product is suitable for laboratory-scale experiments and provides a reliable tool for studying carbohydrate metabolism, enzyme kinetics, and substrate specificity in biochemical and pharmaceutical research.