Learn More
Medchemexpress LLC 4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-glucopyranoside | 37067-30-4 | MFCD00065481 | 99.9% | 379.36 | 50 MILLIGRAM
SDP
Catalog No. 5000352220
Encompass_Preferred
4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-Glucopyranoside is a fluorogenic substrate for N-acetyl-β-D-glucosaminidase. In vitro, the enzyme functionality is assessed by its ability to remove terminal β-glycosidically bound N-acetylglucosamine and N-acetylgalactosamine residues from this substrate.
  • For research use only.
  • Appears as a white to off-white solid.
  • Soluble in DMSO at 50 mg/ml.
Catalog No. 50-003-52220 Supplier Medchemexpress LLC Supplier No. HY137853100MG
4-Methylumbelliferyl-2-acetamido-2-deoxy-β-D-Glucopyranoside is a fluorogenic substrate for N-acetyl-β-D-glucosaminidase. In vitro, the enzyme functionality is assessed by its ability to remove terminal β-glycosidically bound N-acetylglucosamine and N-acetylgalactosamine residues from this substrate.
  • For research use only.
  • Appears as a white to off-white solid.
  • Soluble in DMSO at 50 mg/ml.