Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylumbelliferyl-2-Acetamido-2-Deoxy-?-D-Glucopyranoside, MP Biomedicals™

Catalog No. MP215168183 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215168183 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215168183 Supplier MP Biomedicals, Inc Supplier No. 0215168183
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Substrate for the fluorimetric, phosphorimetric and spectrophotometric assay of N-acetyl-β-D-glucosaminidase and β-N-acetyl-D-hexosaminidase.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37067-30-4
Molecular Formula C18H21NO8
Molecular Weight (g/mol) 379.37
MDL Number MFCD00065481
InChI Key QCTHLCFVVACBSA-XWEABGILSA-N
Synonym 4-mu-glcnac, 4-methylumbelliferyl-n-acetyl-beta-d-glucosaminide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-methyl-2-oxo-2h-chromen-7-yl oxy tetrahydro-2h-pyran-3-yl acetamide, 4-methylumbelliferyl-2-acetamido-2-deoxy-beta-d-glucopyranoside, 4-methylumbelliferyl n-acetyl-, a-d-glucosaminide, 4-methylumbelliferyl n-acetyl-beta-d-glucosaminide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-methyl-2-oxochromen-7-yl oxy oxan-3-yl acetamide, glcnac-b-4mu, 4-methylumbelliferyl 2-acetamido-2-deoxy-, a-d-glucopyranoside, 4-methylumbelliferyl 2-acetamido-2-deoxy-b-d-glucopyranoside
PubChem CID 2733787
IUPAC Name N-[(3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]oxan-3-yl]acetamide
SMILES CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)OC1OC1=CC=C2C(C)=CC(=O)OC2=C1

Specifications

Melting Point 212°C to 222°C
Quantity 250 mg
Solubility Information Soluble in methanol, warm water (100mg/65ml) and DMF (20mg/mL)
Formula Weight 379.4
Physical Form Powder
Recommended Storage 0°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.