Learn More
Medchemexpress LLC 4-Methylumbelliferyl 25mg | 18323-58-5 | 440.61 | C28H40O4 | 25 MG
SDP
Catalog No. 5000308362
Encompass_Preferred
4-Methylumbelliferyl oleate is a fluorogenic substrate used to measure lipase activity; enzymatic hydrolysis liberates 4-methylumbelliferone, a fluorescent reporter (Ex/Em = 320/450 nm). The material is supplied as a 25 mg vial, is soluble in DMSO, and should be stored protected from light at -20°C.
Catalog No. 50-003-08362 Supplier Medchemexpress LLC Supplier No. HY11709525MG
4-Methylumbelliferyl oleate is a fluorogenic substrate used to measure lipase activity; enzymatic hydrolysis liberates 4-methylumbelliferone, a fluorescent reporter (Ex/Em = 320/450 nm). The material is supplied as a 25 mg vial, is soluble in DMSO, and should be stored protected from light at -20°C.