Learn More
AdipoGen 4-Methylumbelliferyl acetate
SDP
Catalog No. 502560582
Encompass_Preferred
Chemical. CAS 2747-05-9. Formula C12H10O4. MW 218.21. Synthetic. Fluorogenic substrate for esterases, broadly applied to various types of esterases, including carboxyesterases. Measurement of intracellular pH in rat proximal convoluted tubule.
Catalog No. 50-256-0582 Supplier AdipoGen Supplier No. CDXM0010G100
Chemical. CAS 2747-05-9. Formula C12H10O4. MW 218.21. Synthetic. Fluorogenic substrate for esterases, broadly applied to various types of esterases, including carboxyesterases. Measurement of intracellular pH in rat proximal convoluted tubule.