Learn More
Medchemexpress LLC 4-methylumbelliferyl alpha-D-glucopyranoside | 17833-43-1 | MFCD00067661 | >99.0% | C16H18O8 | 10MG
SDP
Catalog No. 5000258996
Encompass_Preferred
4-Methylumbelliferyl-α-D-glucopyranoside is a fluorogenic substrate for α-glucosidase assays. Enzymatic cleavage releases the fluorescent 4-methylumbelliferone (4-MU) moiety, enabling sensitive quantification of enzyme activity in biochemical research and diagnostic applications. Supplied as a high-purity crystalline powder suitable for analytical use.
  • Fluorogenic substrate for α-glucosidase assays.
  • Releases 4-methylumbelliferone upon hydrolysis for fluorescence detection.
  • High purity (>99%) for reliable assay performance.
  • Stable solid form for convenient storage and handling.
  • Applicable to enzyme kinetics, screening, and diagnostic assays.
Catalog No. 50-002-58996 Supplier Medchemexpress LLC Supplier No. HY13787310MG
4-Methylumbelliferyl-α-D-glucopyranoside is a fluorogenic substrate for α-glucosidase assays. Enzymatic cleavage releases the fluorescent 4-methylumbelliferone (4-MU) moiety, enabling sensitive quantification of enzyme activity in biochemical research and diagnostic applications. Supplied as a high-purity crystalline powder suitable for analytical use.
  • Fluorogenic substrate for α-glucosidase assays.
  • Releases 4-methylumbelliferone upon hydrolysis for fluorescence detection.
  • High purity (>99%) for reliable assay performance.
  • Stable solid form for convenient storage and handling.
  • Applicable to enzyme kinetics, screening, and diagnostic assays.