Learn More
Medchemexpress LLC 4-methylumbelliferyl alpha-L-arabinofuranoside | 77471-44-4 | MFCD00057308 | 99.9% | C15H16O7 | 10MG
SDP
Catalog No. 5000259652
Encompass_Preferred
4-Methylumbelliferyl alpha-L-arabinofuranoside is a fluorogenic glycosidase substrate used to measure α-L-arabinofuranosidase activity. Enzymatic cleavage releases the fluorescent reporter 4-methylumbelliferone, enabling sensitive fluorometric assays for enzyme kinetics, inhibitor screening, and biochemical studies.
  • Fluorogenic substrate for α-L-arabinofuranosidase assays.
  • Releases 4-methylumbelliferone for high-sensitivity fluorometric detection.
  • Suitable for enzyme kinetics, inhibitor screening, and activity profiling.
  • High purity improves assay reproducibility and signal-to-noise ratio.
  • Available in small, research-scale quantities for laboratory use.
Catalog No. 50-002-59652 Supplier Medchemexpress LLC Supplier No. HY13749010MG
4-Methylumbelliferyl alpha-L-arabinofuranoside is a fluorogenic glycosidase substrate used to measure α-L-arabinofuranosidase activity. Enzymatic cleavage releases the fluorescent reporter 4-methylumbelliferone, enabling sensitive fluorometric assays for enzyme kinetics, inhibitor screening, and biochemical studies.
  • Fluorogenic substrate for α-L-arabinofuranosidase assays.
  • Releases 4-methylumbelliferone for high-sensitivity fluorometric detection.
  • Suitable for enzyme kinetics, inhibitor screening, and activity profiling.
  • High purity improves assay reproducibility and signal-to-noise ratio.
  • Available in small, research-scale quantities for laboratory use.