Learn More
Alkali Scientific 4 Methylumbelliferyl B D glucopyranoside Monohydrate [Methylumbelliferyl B D glucoside], 10 G
SDP
Catalog No. 5000660125
Encompass_Preferred
Methylumbelliferyl β-D Glucopyranoside Monohydrate, 10 g is a biochemical reagent used for the study of glucosidase enzyme activity. This substrate is hydrolyzed by glucosidases to release a fluorescent product, making it an essential tool for enzyme assays, diagnostic tests, and research into metabolic processes. The high purity and sensitivity of this reagent make it ideal for use in microbiology, clinical diagnostics, and genetic research, where detecting small quantities of enzyme activity is crucial. It is particularly useful in enzyme kinetics studies and applications requiring accurate and high-sensitivity measurements of glucosidase activity.
Catalog No. 50-006-60125 Supplier Alkali Scientific Supplier No. OPY003CP
Methylumbelliferyl β-D Glucopyranoside Monohydrate, 10 g is a biochemical reagent used for the study of glucosidase enzyme activity. This substrate is hydrolyzed by glucosidases to release a fluorescent product, making it an essential tool for enzyme assays, diagnostic tests, and research into metabolic processes. The high purity and sensitivity of this reagent make it ideal for use in microbiology, clinical diagnostics, and genetic research, where detecting small quantities of enzyme activity is crucial. It is particularly useful in enzyme kinetics studies and applications requiring accurate and high-sensitivity measurements of glucosidase activity.