Learn More
TARGETMOL CHEMICALS INC 4-Methylumbelliferyl-beta-D-xy
SDP
Catalog No. 502879890
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 4-Methylumbelliferyl-beta-D-xylopyranoside (4-Methylumbelliferyl-beta-D-xyloside) exhibits bursting steady-state kinetics in biological experiments, which supports the conclusion of rapid hydrolysis of the glycosidic bond of the substrate 4-Methylumbelliferyl-beta-D-xylopyranoside 4-Methylumbelliferyl-beta-D-xylopyranoside can be used for beta- xylosidase activity studies. Purity 99.46%
Catalog No. 50-287-9890 Supplier TARGETMOL CHEMICALS INC Supplier No. T67685500MG
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 4-Methylumbelliferyl-beta-D-xylopyranoside (4-Methylumbelliferyl-beta-D-xyloside) exhibits bursting steady-state kinetics in biological experiments, which supports the conclusion of rapid hydrolysis of the glycosidic bond of the substrate 4-Methylumbelliferyl-beta-D-xylopyranoside 4-Methylumbelliferyl-beta-D-xylopyranoside can be used for beta- xylosidase activity studies. Purity 99.46%