Learn More
AdipoGen 4-Methylumbelliferyl butyrate
SDP
Catalog No. 502560579
Encompass_Preferred
Chemical. CAS 17695-46-4. Formula C14H14O4. MW 246.26. Synthetic. Fluorogenic substrate for butyrate esterase.
Catalog No. 50-256-0579 Supplier AdipoGen Supplier No. CDXM0011G125
Chemical. CAS 17695-46-4. Formula C14H14O4. MW 246.26. Synthetic. Fluorogenic substrate for butyrate esterase.