Learn More
Abcam 4-Methylumbelliferyl butyrate, Fluorogenic butyrate esterase substrate, 5G
Catalog No. 502705177
Encompass_Preferred
MW 246.26 g/mol, Purity >95%. Fluorogenic substrate for esterases/lipases, such as butyrate esterase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5177 Supplier Abcam Supplier No. AB1454565G
MW 246.26 g/mol, Purity >95%. Fluorogenic substrate for esterases/lipases, such as butyrate esterase.
The product is subject to the following: Abcam Restricted Use Statement