Learn More
Abcam 4-Methylumbelliferyl butyrate, Fluorogenic butyrate esterase substrate, 20G
Catalog No. 502705178
Encompass_Preferred
MW 246.26 g/mol, Purity >95%. Fluorogenic substrate for esterases/lipases, such as butyrate esterase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5178 Supplier Abcam Supplier No. AB14545620G
MW 246.26 g/mol, Purity >95%. Fluorogenic substrate for esterases/lipases, such as butyrate esterase.
The product is subject to the following: Abcam Restricted Use Statement