Learn More
Medchemexpress LLC HY-D0935A 25mg Medchemexpress, 4-Methylumbelliferyl-β-D-glucuronide hydrate CAS:881005-91-0 Purity:>98%
SDP
Catalog No. 501933211
Encompass_Preferred
Medchemexpress, HY-D0935A 25mg 4-Methylumbelliferyl-β-D-glucuronide hydrate CAS:881005-91-0 4-Methylumbelliferyl-β-D-glucuronide hydrate is a fluorogenic substrat (λ ex =362 nm , λ em =445 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3211 Supplier Medchemexpress LLC Supplier No. HYD0935A25MG
Medchemexpress, HY-D0935A 25mg 4-Methylumbelliferyl-β-D-glucuronide hydrate CAS:881005-91-0 4-Methylumbelliferyl-β-D-glucuronide hydrate is a fluorogenic substrat (λ ex =362 nm , λ em =445 nm). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.