Learn More
Medchemexpress LLC 4-methylumbelliferyl β-D-cellobioside | 72626-61-0 | MFCD00063279 | >98.0% | C22H28O13 | 10MG
SDP
Catalog No. 5000259045
Encompass_Preferred
4-Methylumbelliferyl β-D-cellobioside is a fluorogenic glycoside substrate used to measure cellulase and related β-glucosidase activity. Enzymatic hydrolysis liberates 4-methylumbelliferone, a fluorescent reporter commonly quantified in biochemical and enzymatic assays for activity and kinetics studies.
  • Fluorogenic substrate for cellulase and β-glucosidase assays.
  • Releases fluorescent 4-methylumbelliferone upon enzymatic cleavage.
  • High purity (>98.0%) suitable for quantitative assays.
  • Known molecular weight (500.45 g·mol⁻¹) and chemical formula (C22H28O13).
  • Available in small research-scale quantities for laboratory use.
Catalog No. 50-002-59045 Supplier Medchemexpress LLC Supplier No. HY13733310MG
4-Methylumbelliferyl β-D-cellobioside is a fluorogenic glycoside substrate used to measure cellulase and related β-glucosidase activity. Enzymatic hydrolysis liberates 4-methylumbelliferone, a fluorescent reporter commonly quantified in biochemical and enzymatic assays for activity and kinetics studies.
  • Fluorogenic substrate for cellulase and β-glucosidase assays.
  • Releases fluorescent 4-methylumbelliferone upon enzymatic cleavage.
  • High purity (>98.0%) suitable for quantitative assays.
  • Known molecular weight (500.45 g·mol⁻¹) and chemical formula (C22H28O13).
  • Available in small research-scale quantities for laboratory use.