Learn More
Medchemexpress LLC 4-Methylumbelliferyl β-D-cellobioside | 72626-61-0 | MFCD00063279 | 99.4% | 500.45 | 100 MG
SDP
Catalog No. 5000350216
Encompass_Preferred
4-Methylumbelliferyl β-D-cellobioside is a substrate for cellulase. It can be hydrolyzed to produce fluorescent 4-methylumbelliferone, which is used to study the kinetics of cellulases.
  • Substrate for cellulase
  • Produces fluorescent 4-methylumbelliferone
  • Used to study the kinetics of cellulases
Catalog No. 50-003-50216 Supplier Medchemexpress LLC Supplier No. HY137333100MG
4-Methylumbelliferyl β-D-cellobioside is a substrate for cellulase. It can be hydrolyzed to produce fluorescent 4-methylumbelliferone, which is used to study the kinetics of cellulases.
  • Substrate for cellulase
  • Produces fluorescent 4-methylumbelliferone
  • Used to study the kinetics of cellulases