Learn More
Medchemexpress LLC 4-methylumbelliferyl α-D-galactopyranoside | 38597-12-5 | M65400 | 99.8% | 338.31 g/mol | C16H18O8 | 25 MG
SDP
Catalog No. 5000227832
Encompass_Preferred
4-Methylumbelliferyl-α-D-galactopyranoside is a pro-fluorogenic substrate for α-galactosidase that releases fluorescent 4-methylumbelliferone (4-MU) upon enzymatic cleavage, enabling sensitive fluorometric detection in biochemical and enzymatic assays. Supplied as a powder with documented solubility in DMSO and defined storage conditions for stability.
  • Fluorogenic substrate for α-galactosidase assays.
  • Generates fluorescent 4-methylumbelliferone on enzymatic cleavage.
  • High purity (99.8%) suitable for biochemical assays.
  • Soluble in DMSO (≈33.3 mg/mL) for preparation of stock solutions.
  • Stable when stored as powder at -20°C for long-term storage.
Catalog No. 50-002-27832 Supplier Medchemexpress LLC Supplier No. HY11813525MG
4-Methylumbelliferyl-α-D-galactopyranoside is a pro-fluorogenic substrate for α-galactosidase that releases fluorescent 4-methylumbelliferone (4-MU) upon enzymatic cleavage, enabling sensitive fluorometric detection in biochemical and enzymatic assays. Supplied as a powder with documented solubility in DMSO and defined storage conditions for stability.
  • Fluorogenic substrate for α-galactosidase assays.
  • Generates fluorescent 4-methylumbelliferone on enzymatic cleavage.
  • High purity (99.8%) suitable for biochemical assays.
  • Soluble in DMSO (≈33.3 mg/mL) for preparation of stock solutions.
  • Stable when stored as powder at -20°C for long-term storage.