Learn More
Medchemexpress LLC 4-methylumbelliferyl α-D-galactopyranoside | 38597-12-5 | MFCD00063278 | >98.0% | 338.31 g·mol⁻¹ | C16H18O8 | 100 MG
SDP
Catalog No. 5000226866
Encompass_Preferred
4-Methylumbelliferyl-α-D-galactopyranoside is a fluorogenic substrate for α-galactosidase enzymes. Hydrolysis liberates 4-methylumbelliferone, a fluorescent reporter used to quantify enzyme activity in biochemical assays and diagnostics. Supplied as a solid for laboratory use.
  • High purity (≥98%) suitable for enzymatic assays.
  • Generates a fluorescent product with emission maximum ~445-454 nm.
  • pH-dependent excitation maxima for flexible assay conditions (≈330-385 nm).
  • Solid form for straightforward handling and storage.
  • Suitable for α-galactosidase activity measurements and diagnostic workflows.
Catalog No. 50-002-26866 Supplier Medchemexpress LLC Supplier No. HY118135100MG
4-Methylumbelliferyl-α-D-galactopyranoside is a fluorogenic substrate for α-galactosidase enzymes. Hydrolysis liberates 4-methylumbelliferone, a fluorescent reporter used to quantify enzyme activity in biochemical assays and diagnostics. Supplied as a solid for laboratory use.
  • High purity (≥98%) suitable for enzymatic assays.
  • Generates a fluorescent product with emission maximum ~445-454 nm.
  • pH-dependent excitation maxima for flexible assay conditions (≈330-385 nm).
  • Solid form for straightforward handling and storage.
  • Suitable for α-galactosidase activity measurements and diagnostic workflows.