Learn More
Medchemexpress LLC 4-methylumbelliferyl α-D-galactopyranoside | 38597-12-5 | MFCD00063278 | 99.8% | 338.31 g/mol | C16H18O8 | 1 ML
SDP
Catalog No. 5000227834
Encompass_Preferred
4-Methylumbelliferyl-α-D-galactopyranoside is a fluorogenic substrate for α-galactosidase that releases 4-methylumbelliferone, producing a fluorescent signal used to measure enzyme activity. The product is supplied as a 10 mM solution in DMSO (1 mL) and is commonly used in biochemical assays and high-throughput screening.
  • Supplied as 10 mM in DMSO, 1 mL vial.
  • High reported purity (≈99.8%).
  • Generates a strong fluorometric signal upon enzymatic cleavage.
  • Suitable for α-galactosidase activity assays and screening workflows.
  • Available in solution and solid pack sizes for assay flexibility.
Catalog No. 50-002-27834 Supplier Medchemexpress LLC Supplier No. HY11813510MM 1ML
4-Methylumbelliferyl-α-D-galactopyranoside is a fluorogenic substrate for α-galactosidase that releases 4-methylumbelliferone, producing a fluorescent signal used to measure enzyme activity. The product is supplied as a 10 mM solution in DMSO (1 mL) and is commonly used in biochemical assays and high-throughput screening.
  • Supplied as 10 mM in DMSO, 1 mL vial.
  • High reported purity (≈99.8%).
  • Generates a strong fluorometric signal upon enzymatic cleavage.
  • Suitable for α-galactosidase activity assays and screening workflows.
  • Available in solution and solid pack sizes for assay flexibility.